Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006579.2 | NP_006570.1 | 3 | 531 | ||
GenBank mRNA | BC001549.1 | - | 3 | 461 | 3463689 | |
BC001572.1 | - | 3 | 463 | 3453423 | ||
BC046501.1 | - | 3 | 505 | 5106272 | ||
CA488777.1 | - | 3 | 511 | |||
CR456815.1 | - | 2 | 326 | |||
CR542094.1 | - | 2 | 326 | |||
KJ921656.1 | - | 2 | 326 | |||
Z37986.1 | - | 3 | 437 |