Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006876.2 | NP_006867.1 | 2 | 1258 | ||
GenBank mRNA | AF029893.1 | - | 2 | 1192 | ||
AK075354.1 | - | 2 | 1142 | |||
AK098389.1 | - | 2 | 1197 | |||
AK098779.1 | - | 2 | 1142 | |||
AK225182.1 | - | 2 | 1145 | |||
AK293715.1 | - | 3 | 1166 | |||
AK294604.1 | - | 3 | 1076 | |||
BC021965.2 | - | 2 | 1182 | 2988041 |