Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL3RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267713.1 | NP_001254642.1 | 8, 9 | 1103 | ||
NM_002183.3 | NP_002174.1 | 10, 11 | 1337 | |||
XM_005274431.4 | XP_005274488.1 | 10, 11 | 1374 | |||
XM_005274432.1 | XP_005274489.1 | 10, 11 | 1164 | |||
XM_017029491.1 | XP_016884980.1 | 10, 11 | 1372 | |||
GenBank mRNA | AK290568.1 | - | 10, 11 | 1167 | ||
BC035407.1 | - | 10, 11 | 1337 | 5167281 | ||
FJ550347.1 | - | 7, 8 | 754 | |||
M74782.1 | - | 10, 11 | 1134 |