Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMMR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142556.1 | NP_001136028.1 | 5 | 542 | ||
NM_001142557.1 | NP_001136029.1 | 2 | 315 | |||
NM_012484.2 | NP_036616.2 | 5 | 539 | |||
NM_012485.2 | NP_036617.2 | 4 | 494 | |||
GenBank mRNA | AF032862.1 | - | 5 | 393 | ||
AK290577.1 | - | 4 | 335 | |||
AK303616.1 | - | 2 | 193 | |||
AK313603.1 | - | 5 | 460 | |||
AY623426.1 | - | 5 | 365 | |||
BC017793.1 | - | 4 | 363 | |||
BC030505.2 | - | 5 | 449 | |||
BC057235.1 | - | 4 | 464 | |||
BC070281.1 | - | 4 | 370 | |||
BC108904.1 | - | 5 | 394 | 40007903 | ||
DB091289.1 | - | 4 | 494 | |||
U29343.1 | - | 5 | 420 |