Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGAT4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013244.3 | NP_037376.2 | 6, 7 | 769 | ||
XM_005268776.4 | XP_005268833.1 | 5, 6 | 526 | |||
XM_005268779.3 | XP_005268836.1 | 4, 5 | 1294 | |||
XM_011538150.2 | XP_011536452.1 | 5, 6 | 593 | |||
XM_011538151.2 | XP_011536453.1 | 5, 6 | 509 | |||
XM_011538156.2 | XP_011536458.1 | 8, 9 | 1801 | |||
XM_011538157.2 | XP_011536459.1 | 7, 8 | 2460 | |||
XM_017019143.1 | XP_016874632.1 | 5, 6 | 1318 | |||
XM_017019144.1 | XP_016874633.1 | 5, 6 | 499 | |||
XM_017019145.1 | XP_016874634.1 | 6, 7 | 634 | |||
XM_017019146.1 | XP_016874635.1 | 6, 7 | 631 | |||
XM_017019147.1 | XP_016874636.1 | 5, 6 | 609 | |||
XM_017019148.1 | XP_016874637.1 | 7, 8 | 1593 | |||
XM_017019149.1 | XP_016874638.1 | 6, 7 | 1515 | |||
XM_017019150.1 | XP_016874639.1 | 7, 8 | 897 | |||
GenBank mRNA | AK127727.1 | - | 7, 8 | 1508 | ||
BC026068.1 | - | 5, 6 | 533 | |||
BC064141.1 | - | 2, 3 | 310 | 30378187 |