Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307986.1 | NP_001294915.1 | 4 | 1008 | ||
NM_001307987.1 | NP_001294916.1 | 2 | 807 | |||
NM_013305.5 | NP_037437.2 | 3 | 900 | |||
XM_017025715.1 | XP_016881204.1 | 5 | 396 | |||
XM_017025716.1 | XP_016881205.1 | 3 | 541 | |||
GenBank mRNA | AK056270.1 | - | 3 | 900 | ||
AK130126.1 | - | 5 | 455 | |||
AK293608.1 | - | 4 | 648 | |||
AK298881.1 | - | 2 | 448 | |||
AK310764.1 | - | 4 | 649 | |||
BC108910.1 | - | 3 | 450 | 40011279 | ||
BC108911.1 | - | 3 | 450 | 40011281 | ||
CR457037.1 | - | 3 | 239 | |||
U91641.1 | - | 3 | 286 |