Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GNT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014256.3 | NP_055071.2 | 3 | 1187 | ||
XM_011527626.2 | XP_011525928.1 | 3 | 1146 | |||
GenBank mRNA | AB015630.1 | - | 3 | 1157 | ||
AB049585.1 | - | 3 | 1142 | |||
AF293973.1 | - | 2 | 1053 | |||
AJ130847.1 | - | 2 | 1040 | |||
AJ278961.1 | - | 2 | 1046 | |||
AK075434.1 | - | 4 | 737 | |||
AK075466.1 | - | 3 | 1162 | |||
AK225421.1 | - | 3 | 1154 | |||
AK314323.1 | - | 3 | 1158 | |||
AY358955.1 | - | 3 | 1142 | |||
BC066876.1 | - | 3 | 1159 | 6971715 | ||
BC067423.1 | - | 3 | 1138 | 6971716 |