Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289905.1 | NP_001276834.1 | 3 | 323 | ||
NM_014339.6 | NP_055154.3 | 3 | 323 | |||
GenBank mRNA | BC011624.2 | - | 3 | 267 | 4053746 | |
BX334547.2 | - | 3 | 326 | |||
BX410574.2 | - | 3 | 239 | |||
CB044543.1 | - | 2 | 306 | |||
CT841520.1 | - | 3 | 246 | |||
JF305973.1 | - | 3 | 190 | |||
U58917.1 | - | 3 | 222 |