Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128928.1 | NP_001122400.1 | 5 | 992 | ||
NM_002194.3 | NP_002185.1 | 4 | 848 | |||
XM_005246532.1 | XP_005246589.1 | 4 | 629 | |||
GenBank mRNA | AB451298.1 | - | 3 | 292 | ||
AK093560.1 | - | 6 | 950 | |||
BC015496.1 | - | 3 | 358 | 3845956 | ||
BC106006.1 | - | 4 | 392 | 4472604 | ||
CR456840.1 | - | 3 | 292 | |||
CR541707.1 | - | 3 | 292 | |||
KJ535023.1 | - | 3 | 292 | |||
KJ535042.1 | - | 3 | 291 | |||
KU178146.1 | - | 3 | 292 | |||
L08488.1 | - | 4 | 618 |