Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHCR24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014762.3 | NP_055577.1 | 5 | 884 | ||
GenBank mRNA | AB073391.1 | - | 6 | 759 | ||
AF261758.1 | - | 5 | 854 | |||
AK302774.1 | - | 5 | 715 | |||
AK304302.1 | - | 3 | 316 | |||
AK304437.1 | - | 5 | 601 | |||
AK316419.1 | - | 5 | 646 | |||
AK316427.1 | - | 5 | 585 | |||
BC004375.1 | - | 5 | 790 | 3638639 | ||
BC011669.2 | - | 5 | 817 | 4130396 | ||
D13643.3 | - | 5 | 867 |