Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECTIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243286.1 | NP_001230215.1 | 2 | 715 | ||
NM_001243288.1 | NP_001230217.1 | 2 | 639 | |||
NM_015480.2 | NP_056295.1 | 2 | 715 | |||
XM_011512663.1 | XP_011510965.1 | 3 | 779 | |||
XM_017006123.1 | XP_016861612.1 | 3 | 779 | |||
XM_017006124.1 | XP_016861613.1 | 3 | 436 | |||
XM_017006125.1 | XP_016861614.1 | 2 | 639 | |||
XM_017006126.1 | XP_016861615.1 | 2 | 710 | |||
XM_017006127.1 | XP_016861616.1 | 2 | 802 | |||
GenBank mRNA | AF282874.1 | - | 2 | 456 | ||
AK075105.1 | - | 2 | 639 | |||
AL050071.1 | - | 1 | 32 | |||
BC067808.1 | - | 2 | 598 | 5285526 |