Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172477.1 | NP_001165948.1 | 9 | 1294 | ||
NM_001172478.1 | NP_001165949.1 | 8 | 1157 | |||
NM_015713.4 | NP_056528.2 | 9 | 1313 | |||
GenBank mRNA | AB036063.1 | - | 9 | 1313 | ||
AK001965.1 | - | 9 | 1108 | |||
AK304354.1 | - | 9 | 1238 | |||
AL137348.1 | - | 9 | 1115 | |||
BC042468.1 | - | 7 | 871 | |||
BC108261.1 | - | 9 | 1125 | 6143754 | ||
BC117496.1 | - | 9 | 1150 | 40126047 | ||
BC130628.1 | - | 9 | 1150 | 40146659 | ||
BX509409.1 | - | 3 | 317 | |||
CR627376.1 | - | 9 | 1238 |