Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015879.2 | NP_056963.2 | 3 | 683 | ||
GenBank mRNA | AB208996.1 | - | 3 | 687 | ||
AF003092.2 | - | 3 | 398 | |||
AF004668.2 | - | 3 | 2349 | |||
AK289517.1 | - | 3 | 461 | |||
AK313047.1 | - | 3 | 409 | |||
BC074909.2 | - | 3 | 374 | 30915256 |