Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204830.1 | NP_001191759.1 | 3 | 611 | ||
NM_015929.3 | NP_057013.1 | 3 | 413 | |||
NM_145197.2 | NP_660198.1 | 4 | 541 | |||
NM_145198.2 | NP_660199.1 | 4 | 529 | |||
NM_145199.2 | NP_660200.1 | 2 | 287 | |||
NR_037935.1 | 4 | 674 | ||||
NR_037936.1 | 3 | 350 | ||||
GenBank mRNA | AB017566.1 | - | 3 | 413 | ||
BC007001.1 | - | 3 | 333 | 3961227 | ||
BC009772.1 | - | 3 | 354 | 4091817 | ||
BC070145.1 | - | 3 | 391 | 4671058 | ||
BG717409.1 | - | 3 | 589 | |||
BQ422048.1 | - | 3 | 322 | |||
BQ432795.1 | - | 4 | 628 |