Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016059.4 | NP_057143.1 | 2 | 338 | ||
GenBank mRNA | AF151882.1 | - | 2 | 313 | ||
AK026636.1 | - | 2 | 140 | |||
AY359032.1 | - | 2 | 315 | |||
BC003048.1 | - | 2 | 290 | 3543388 | ||
BU195579.1 | - | 2 | 339 |