Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB5R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302826.1 | NP_001289755.1 | 6 | 1023 | ||
NM_001302827.1 | NP_001289756.1 | 5 | 497 | |||
NM_016229.4 | NP_057313.2 | 6 | 677 | |||
NR_126508.1 | 6 | 677 | ||||
XM_005252975.4 | XP_005253032.1 | 6 | 629 | |||
XM_006718251.2 | XP_006718314.1 | 6 | 654 | |||
XM_011520184.2 | XP_011518486.1 | 6 | 844 | |||
XM_011520185.2 | XP_011518487.1 | 8 | 943 | |||
XM_017017920.1 | XP_016873409.1 | 8 | 937 | |||
XM_017017921.1 | XP_016873410.1 | 8 | 939 | |||
XM_017017922.1 | XP_016873411.1 | 6 | 571 | |||
GenBank mRNA | AB209000.1 | - | 2 | 4713 | ||
AF169802.1 | - | 6 | 580 | |||
AK290102.1 | - | 6 | 996 | |||
AK308543.1 | - | 6 | 1180 | |||
AL133582.1 | - | 5 | 413 | |||
AY665398.1 | - | 5 | 431 | |||
BC001346.1 | - | 6 | 551 | 3451742 | ||
BG495762.1 | - | 6 | 570 | |||
BQ431043.1 | - | 5 | 435 |