Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166663.1 | NP_001160135.1 | 9 | 1519 | ||
NM_001166664.1 | NP_001160136.1 | 8 | 1228 | |||
NM_016382.3 | NP_057466.1 | 9 | 1504 | |||
XM_011509623.2 | XP_011507925.1 | 7 | 904 | |||
GenBank mRNA | AF107761.2 | - | 9 | 1387 | ||
AF117711.1 | - | 9 | 1458 | |||
AF145782.1 | - | 9 | 1279 | |||
BC028073.1 | - | 9 | 1504 | 5213370 | ||
BC041607.1 | - | 9 | 1306 | |||
BC053985.1 | - | 9 | 1290 | 5484877 |