Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017421.3 | NP_059117.3 | 6 | 864 | ||
XM_005267018.4 | XP_005267075.1 | 5 | 637 | |||
XM_006715500.3 | XP_006715563.1 | 7 | 1003 | |||
XM_011535898.2 | XP_011534200.1 | 6 | 761 | |||
XM_011535899.1 | XP_011534201.1 | 5 | 692 | |||
XM_017010933.1 | XP_016866422.1 | 7 | 861 | |||
GenBank mRNA | AF193016.1 | - | 6 | 801 | ||
AF351615.1 | - | 6 | 869 | |||
AK056955.1 | - | 6 | 862 | |||
AL136726.1 | - | 6 | 862 | |||
BC001534.1 | - | 6 | 835 | |||
BC015634.2 | - | 6 | 825 | |||
BC063463.1 | - | 6 | 840 | 5196692 |