Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEPROT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198681.1 | NP_001185610.1 | 5 | 536 | ||
NM_017526.4 | NP_059996.1 | 4 | 672 | |||
GenBank mRNA | AK074841.1 | - | 4 | 433 | ||
AK130096.1 | - | 1 | 966 | |||
AK309370.1 | - | 5 | 536 | |||
BC011027.2 | - | 3 | 380 | |||
BC056250.1 | - | 4 | 417 | 4328021 | ||
BI715133.1 | - | 3 | 349 | |||
Y12670.1 | - | 4 | 465 |