Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BHMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178005.1 | NP_001171476.1 | 7 | 1382 | ||
NM_017614.4 | NP_060084.2 | 8 | 1574 | |||
GenBank mRNA | AF257473.1 | - | 8 | 1549 | ||
AK000008.1 | - | 7 | 1486 | |||
AK298298.1 | - | 7 | 1339 | |||
BC020665.1 | - | 8 | 1529 | 4722608 |