Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312691.1 | NP_001299620.1 | 2 | 235 | ||
NM_017646.5 | NP_060116.2 | 2 | 235 | |||
NR_132401.1 | 2 | 235 | ||||
NR_132402.1 | 2 | 235 | ||||
NR_132403.1 | 2 | 235 | ||||
NR_132404.1 | 2 | 235 | ||||
NR_132405.1 | 2 | 235 | ||||
NR_132406.1 | 2 | 235 | ||||
NR_132415.1 | 2 | 235 | ||||
XM_005270954.2 | XP_005271011.1 | 2 | 471 | |||
XM_006710706.1 | XP_006710769.1 | 2 | 228 | |||
GenBank mRNA | AF074918.1 | - | 2 | 229 | ||
AK223478.1 | - | 2 | 228 | |||
AK296453.1 | - | 2 | 220 | |||
AK304759.1 | - | 2 | 228 | |||
AK307910.1 | - | 2 | 220 | |||
AY052768.1 | - | 2 | 197 | |||
AY303390.1 | - | 2 | 229 | |||
AY702933.1 | - | 2 | 158 | |||
AY702934.1 | - | 2 | 158 | |||
AY702935.1 | - | 2 | 158 | |||
AY702936.1 | - | 2 | 158 | |||
AY702937.1 | - | 2 | 158 | |||
AY702938.1 | - | 2 | 158 | |||
AY702939.1 | - | 2 | 158 | |||
AY702940.1 | - | 2 | 158 | |||
BC107569.1 | - | 2 | 234 | 4594069 | ||
BC128155.1 | - | 2 | 229 | 40112695 | ||
BC128156.1 | - | 2 | 229 | 40112698 | ||
DA031689.1 | - | 2 | 230 |