Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018010.3 | NP_060480.1 | 2, 3 | 619 | ||
GenBank mRNA | AB014762.1 | - | 2, 3 | 371 | ||
AF139576.1 | - | 2, 3 | 449 | |||
AF245220.1 | - | 2, 3 | 513 | |||
AK001009.1 | - | 2, 3 | 458 | |||
AK298854.1 | - | 2, 3 | 517 | |||
BC011899.2 | - | 2, 3 | 468 | 4053749 | ||
CR457251.1 | - | 2, 3 | 371 |