Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMLHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184797.1 | NP_001171726.1 | 3 | 426 | ||
NM_018196.3 | NP_060666.1 | 3 | 426 | |||
XM_011531182.2 | XP_011529484.1 | 3 | 294 | |||
XM_017029620.1 | XP_016885109.1 | 3 | 426 | |||
GenBank mRNA | AF373407.1 | - | 3 | 297 | ||
AK001589.1 | - | 3 | 401 | |||
AK291694.1 | - | 3 | 364 | |||
AK304830.1 | - | 1 | 60 | |||
AK310667.1 | - | 1 | 61 | |||
BC025269.1 | - | 3 | 342 | 3689631 | ||
CR457265.1 | - | 2 | 255 | |||
DA323339.1 | - | 3 | 422 | |||
DB174485.1 | - | 3 | 430 |