Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034023.1 | NP_001029195.1 | 4 | 362 | ||
NM_001034024.1 | NP_001029196.1 | 4 | 322 | |||
NM_019100.4 | NP_061973.1 | 3 | 543 | |||
XM_005271037.4 | XP_005271094.1 | 3 | 533 | |||
XM_005271039.1 | XP_005271096.1 | 4 | 433 | |||
XM_005271040.1 | XP_005271097.1 | 4 | 310 | |||
XM_006710771.3 | XP_006710834.1 | 2 | 212 | |||
XM_011541784.2 | XP_011540086.1 | 4 | 367 | |||
XM_011541785.2 | XP_011540087.1 | 4 | 387 | |||
XM_017001807.1 | XP_016857296.1 | 4 | 418 | |||
XM_017001808.1 | XP_016857297.1 | 4 | 421 | |||
GenBank mRNA | AB037846.1 | - | 4 | 327 | ||
AF265228.1 | - | 3 | 242 | |||
AK021605.1 | - | 4 | 306 | |||
AK289366.1 | - | 4 | 345 | |||
AK293598.1 | - | 4 | 353 | |||
AK300217.1 | - | 4 | 320 | |||
AK300372.1 | - | 4 | 312 | |||
AK300442.1 | - | 5 | 428 | |||
AL136657.1 | - | 4 | 324 | |||
AL137200.1 | - | 4 | 326 | |||
BC002855.2 | - | 4 | 311 | 3635724 | ||
BC008053.2 | - | 3 | 505 | 3834056 | ||
CA842195.1 | - | 4 | 354 |