Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019109.4 | NP_061982.3 | 5 | 619 | ||
XM_011522565.1 | XP_011520867.1 | 5 | 426 | |||
XM_017023457.1 | XP_016878946.1 | 5 | 619 | |||
XM_017023458.1 | XP_016878947.1 | 5 | 741 | |||
GenBank mRNA | AB019038.1 | - | 5 | 593 | ||
AK075373.1 | - | 5 | 609 | |||
AK298144.1 | - | 5 | 426 | |||
AK311475.1 | - | 5 | 744 | |||
AY359073.1 | - | 5 | 603 | |||
BC004402.1 | - | 5 | 586 | 3510605 | ||
BC031095.1 | - | 5 | 598 | 5276692 | ||
BC031873.2 | - | 6 | 1345 |