Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IVD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159508.1 | NP_001152980.1 | 4 | 769 | ||
NM_002225.3 | NP_002216.2 | 5 | 859 | |||
XM_005254350.2 | XP_005254407.1 | 5 | 525 | |||
XM_006720492.2 | XP_006720555.1 | 5 | 525 | |||
XM_006720495.2 | XP_006720558.1 | 5 | 525 | |||
XM_017022149.1 | XP_016877638.1 | 5 | 644 | |||
XM_017022150.1 | XP_016877639.1 | 5 | 643 | |||
XM_017022151.1 | XP_016877640.1 | 5 | 643 | |||
XM_017022152.1 | XP_016877641.1 | 5 | 640 | |||
XM_017022153.1 | XP_016877642.1 | 5 | 642 | |||
XM_017022154.1 | XP_016877643.1 | 5 | 781 | |||
XM_017022155.1 | XP_016877644.1 | 5 | 641 | |||
XM_017022156.1 | XP_016877645.1 | 5 | 926 | |||
XM_017022157.1 | XP_016877646.1 | 5 | 643 | |||
XM_017022158.1 | XP_016877647.1 | 5 | 643 | |||
GenBank mRNA | AK122922.1 | - | 4 | 459 | ||
AK315296.1 | - | 5 | 525 | |||
BC017202.2 | - | 5 | 527 | 3627787 | ||
BT007145.1 | - | 5 | 516 | |||
KU178150.1 | - | 5 | 516 | |||
KU178151.1 | - | 4 | 426 | |||
M34192.1 | - | 5 | 531 |