Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACKR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020311.2 | NP_064707.1 | 2 | 1384 | ||
XM_005246097.2 | XP_005246154.1 | 2 | 1280 | |||
XM_005246098.3 | XP_005246155.1 | 3 | 1522 | |||
XM_017004516.1 | XP_016860005.1 | 2 | 1275 | |||
GenBank mRNA | AF030297.1 | - | 2 | 1232 | ||
AK291659.1 | - | 2 | 1296 | |||
BC008459.1 | - | 1 | 781 | |||
BC036661.2 | - | 4 | 1581 | 5267661 | ||
U67784.1 | - | 1 | 990 |