Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CROT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143935.1 | NP_001137407.1 | 19 | 2195 | ||
NM_021151.3 | NP_066974.2 | 18 | 2111 | |||
XM_017012370.1 | XP_016867859.1 | 19 | 2185 | |||
GenBank mRNA | AF052187.1 | - | 4 | 382 | ||
AF073770.1 | - | 18 | 2002 | |||
AF168793.1 | - | 18 | 2002 | |||
AK127901.1 | - | 19 | 2151 | |||
AK300114.1 | - | 13 | 1441 | |||
BC039004.1 | - | 18 | 2051 | 5242962 |