Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SQRDL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271213.1 | NP_001258142.1 | 3 | 327 | ||
NM_021199.3 | NP_067022.1 | 2 | 520 | |||
GenBank mRNA | AF042284.1 | - | 2 | 210 | ||
AF118085.1 | - | 1 | 41 | |||
AF151802.1 | - | 2 | 204 | |||
AK130140.1 | - | 3 | 327 | |||
AK222463.1 | - | 2 | 219 | |||
BC016836.1 | - | 2 | 175 | 3884024 | ||
DA821218.1 | - | 2 | 520 |