Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL22RA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021258.3 | NP_067081.2 | 2, 3 | 372 | ||
XM_011541882.1 | XP_011540184.1 | 2, 3 | 266 | |||
GenBank mRNA | AF286095.1 | - | 2, 3 | 192 | ||
AK226001.1 | - | 2, 3 | 206 | |||
AK292204.1 | - | 2, 3 | 267 | |||
AK308522.1 | - | 2, 3 | 215 | |||
AK313971.1 | - | 2, 3 | 372 | |||
BC029273.1 | - | 2, 3 | 208 | 4347189 |