Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL21R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021798.3 | NP_068570.1 | 4 | 367 | ||
NM_181078.2 | NP_851564.1 | 4 | 720 | |||
NM_181079.4 | NP_851565.4 | 5 | 861 | |||
XM_011545857.2 | XP_011544159.1 | 5 | 897 | |||
XM_011545858.2 | XP_011544160.1 | 5 | 861 | |||
XM_017023257.1 | XP_016878746.1 | 5 | 850 | |||
GenBank mRNA | AF254067.1 | - | 4 | 315 | ||
AF269133.1 | - | 4 | 383 | |||
AK292663.1 | - | 4 | 720 | |||
AK312825.1 | - | 4 | 709 | |||
AY358826.1 | - | 4 | 350 | |||
BC004348.1 | - | 4 | 332 | 3634520 | ||
BQ215949.1 | - | 4 | 380 | |||
BX401767.2 | - | 4 | 480 | |||
DB115361.1 | - | 5 | 508 |