Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGFRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004356.2 | NP_001004356.1 | 6 | 1105 | ||
NM_001004358.1 | NP_001004358.1 | 6 | 988 | |||
NM_021923.3 | NP_068742.2 | 5 | 978 | |||
XM_011513486.1 | XP_011511788.1 | 6 | 1109 | |||
GenBank mRNA | AF279689.1 | - | 6 | 988 | ||
AF312678.1 | - | 1105 | ||||
AJ277437.1 | - | 5 | 978 | |||
AK172829.1 | - | 5 | 962 | |||
AK225486.1 | - | 5 | 949 | |||
AK314365.1 | - | 6 | 1106 | |||
AY358303.1 | - | 1105 | ||||
BC015228.1 | - | 2 | 389 | |||
BC036769.1 | - | 6 | 1117 | 5744684 |