Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022336.3 | NP_071731.1 | 3 | 515 | ||
XM_006712204.1 | XP_006712267.1 | 3 | 399 | |||
XM_011510502.2 | XP_011508804.2 | 3 | 278 | |||
XM_011510503.2 | XP_011508805.2 | 3 | 271 | |||
GenBank mRNA | AF130988.1 | - | 3 | 516 | ||
AK296936.1 | - | 3 | 309 | |||
AK313781.1 | - | 3 | 399 | |||
BC093870.1 | - | 2 | 107 | 7939715 | ||
BC093872.1 | - | 2 | 107 | 7939717 | ||
CR749352.1 | - | 4 | 528 |