Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002264.3 | NP_002255.3 | 3 | 403 | ||
NR_026698.1 | 3 | 403 | ||||
XM_005247437.3 | XP_005247494.1 | 3 | 285 | |||
XM_011512795.2 | XP_011511097.1 | 4 | 337 | |||
XM_011512797.2 | XP_011511099.1 | 2 | 220 | |||
XM_017006363.1 | XP_016861852.1 | 3 | 401 | |||
XM_017006364.1 | XP_016861853.1 | 3 | 280 | |||
GenBank mRNA | S75295.1 | - | 3 | 261 | ||
U20620.1 | - | 1 | 68 |