Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007027.2 | NP_001007028.1 | 4 | 493 | ||
NM_024079.4 | NP_076984.2 | 4 | 493 | |||
XM_005274247.3 | XP_005274304.1 | 4 | 1254 | |||
XM_011545252.2 | XP_011543554.1 | 4 | 503 | |||
XM_017018274.1 | XP_016873763.1 | 4 | 1241 | |||
GenBank mRNA | AJ224875.1 | - | 4 | 447 | ||
AK075184.1 | - | 4 | 433 | |||
AK075194.1 | - | 4 | 448 | |||
AK098614.1 | - | 4 | 493 | |||
AK309050.1 | - | 4 | 462 | |||
BC001133.2 | - | 4 | 476 | 2966827 | ||
KU178764.1 | - | 4 | 428 |