Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007027.2 | NP_001007028.1 | 12 | 1386 | ||
NM_024079.4 | NP_076984.2 | 12 | 1386 | |||
XM_005274247.3 | XP_005274304.1 | 12 | 2147 | |||
XM_011545252.2 | XP_011543554.1 | 12 | 1396 | |||
XM_017018274.1 | XP_016873763.1 | 12 | 2134 | |||
GenBank mRNA | AA375954.1 | - | 1 | 30 | ||
AJ224875.1 | - | 12 | 1340 | |||
AK075184.1 | - | 12 | 1326 | |||
AK075194.1 | - | 12 | 1341 | |||
AK098614.1 | - | 12 | 1386 | |||
BC001133.2 | - | 12 | 1369 | 2966827 | ||
KU178764.1 | - | 12 | 1321 | |||
KU178765.1 | - | 4 | 378 |