Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320611.1 | NP_001307540.1 | 4 | 878 | ||
NM_002266.3 | NP_002257.1 | 4 | 520 | |||
GenBank mRNA | AK024833.1 | - | 4 | 388 | ||
AK058111.1 | - | 4 | 389 | |||
AK291954.1 | - | 4 | 387 | |||
BC005978.1 | - | 4 | 387 | 4105058 | ||
BC013257.1 | - | 4 | 383 | |||
BC034443.1 | - | 4 | 346 | |||
BC053343.1 | - | 4 | 376 | 4450470 | ||
BC067848.1 | - | 4 | 619 | 4825235 | ||
BC146905.1 | - | 4 | 374 | 9056833 | ||
BT006665.1 | - | 3 | 258 | |||
DC388846.1 | - | 4 | 520 | |||
U09559.1 | - | 4 | 390 | |||
U28386.1 | - | 4 | 390 |