Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSSK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052841.3 | NP_443073.1 | 2 | 665 | ||
GenBank mRNA | AF296450.1 | - | 2 | 327 | ||
BC035354.1 | - | 2 | 729 | 5171736 |