Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024642.4 | NP_078918.3 | 8 | 1365 | ||
XM_006717287.1 | XP_006717350.1 | 8 | 1181 | |||
XM_011519018.2 | XP_011517320.1 | 9 | 2037 | |||
XM_011519020.1 | XP_011517322.1 | 8 | 1213 | |||
XM_017015133.1 | XP_016870622.1 | 9 | 1125 | |||
GenBank mRNA | AB040673.1 | - | 8 | 1365 | ||
AB078146.1 | - | 8 | 1365 | |||
AJ132365.2 | - | 8 | 1365 | |||
AJ505963.1 | - | 2 | 144 | |||
AK024865.1 | - | 7 | 938 | |||
BC013945.1 | - | 5 | 530 |