Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYBRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127383.1 | NP_001120855.1 | 2 | 446 | ||
NM_001256909.1 | NP_001243838.1 | 3 | 317 | |||
NM_024843.3 | NP_079119.3 | 3 | 655 | |||
GenBank mRNA | AK128495.1 | - | 2 | 325 | ||
AK291731.1 | - | 3 | 505 | |||
AK301485.1 | - | 3 | 317 | |||
AK316588.1 | - | 3 | 534 | |||
AL136693.1 | - | 3 | 538 | |||
BC065290.1 | - | 3 | 490 | 6063145 | ||
CX866825.1 | - | 3 | 546 |