Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYBRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127383.1 | NP_001120855.1 | 3 | 581 | ||
NM_001256909.1 | NP_001243838.1 | 4 | 452 | |||
NM_024843.3 | NP_079119.3 | 4 | 790 | |||
GenBank mRNA | AK027115.1 | - | 4 | 632 | ||
AK128495.1 | - | 3 | 460 | |||
AK225459.1 | - | 4 | 632 | |||
AK291731.1 | - | 4 | 640 | |||
AK301485.1 | - | 4 | 452 | |||
AK316588.1 | - | 4 | 669 | |||
AL136693.1 | - | 4 | 673 | |||
BC065290.1 | - | 4 | 625 | 6063145 | ||
CX866825.1 | - | 4 | 681 |