Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025163.3 | NP_079439.2 | 3 | 1695 | ||
XM_011513190.1 | XP_011511492.1 | 4 | 2286 | |||
XM_011513192.2 | XP_011511494.1 | 4 | 1812 | |||
XM_017007240.1 | XP_016862729.1 | 3 | 2653 | |||
XM_017007241.1 | XP_016862730.1 | 4 | 4030 | |||
XM_017007242.1 | XP_016862731.1 | 3 | 3722 | |||
XM_017007243.1 | XP_016862732.1 | 3 | 1614 | |||
GenBank mRNA | AK022830.1 | - | 3 | 1657 | ||
AK296867.1 | - | 2 | 1297 | |||
BC018804.1 | - | 3 | 1576 | |||
BC044640.1 | - | 3 | 1577 | 4871581 |