Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GNT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030765.2 | NP_110392.1 | 3 | 458 | ||
XM_011538732.1 | XP_011537034.1 | 2 | 295 | |||
GenBank mRNA | AB049586.1 | - | 2 | 328 | ||
AJ278962.1 | - | 1 | 165 | |||
AK095746.1 | - | 2 | 295 | |||
AL834452.1 | - | 2 | 272 | |||
AY358940.1 | - | 3 | 596 | |||
BC031399.1 | - | 3 | 425 | 5197310 |