Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GALNAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030965.2 | NP_112227.1 | 3 | 828 | ||
GenBank mRNA | AJ507292.1 | - | 3 | 572 | ||
AK056241.1 | - | 3 | 795 | |||
AK095758.1 | - | 4 | 718 | |||
AK295019.1 | - | 4 | 671 | |||
BC001201.1 | - | 3 | 735 | 3356535 | ||
KF548000.1 | - | 1 | 299 |