Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073125.1 | 9 | 1107 | |||
NR_073126.1 | 9 | 1021 | ||||
NR_073127.1 | 7 | 936 | ||||
XM_011519720.2 | XP_011518022.1 | 9 | 1386 | |||
GenBank mRNA | AB040820.1 | - | 7 | 817 | ||
AB055603.1 | - | 9 | 959 | |||
AK074211.1 | - | 9 | 1021 | |||
BC002862.2 | - | 9 | 1050 | 3941289 | ||
BM724648.1 | - | 2 | 197 |