Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032472.3 | NP_115861.1 | 7 | 711 | ||
NM_130906.2 | NP_570981.1 | 7 | 699 | |||
XM_005246651.3 | XP_005246708.1 | 7 | 630 | |||
XM_005246652.4 | XP_005246709.1 | 7 | 618 | |||
XM_011511358.2 | XP_011509660.1 | 7 | 804 | |||
XM_017004352.1 | XP_016859841.1 | 7 | 821 | |||
XM_017004353.1 | XP_016859842.1 | 7 | 560 | |||
XM_017004354.1 | XP_016859843.1 | 7 | 994 | |||
XM_017004355.1 | XP_016859844.1 | 7 | 1018 | |||
XM_017004356.1 | XP_016859845.1 | 7 | 982 | |||
XM_017004357.1 | XP_016859846.1 | 8 | 901 | |||
XM_017004358.1 | XP_016859847.1 | 8 | 724 | |||
XM_017004359.1 | XP_016859848.1 | 8 | 908 | |||
XM_017004360.1 | XP_016859849.1 | 6 | 721 | |||
GenBank mRNA | AF146799.1 | - | 7 | 568 | ||
AF251049.1 | - | 7 | 711 | |||
AF271652.1 | - | 7 | 634 | |||
AF414127.1 | - | 6 | 635 | |||
AY257468.1 | - | 6 | 755 | |||
BC007693.2 | - | 7 | 1140 | 3956681 | ||
CB961908.1 | - | 7 | 754 | |||
KU178646.1 | - | 5 | 458 | |||
KU178647.1 | - | 6 | 511 | |||
KU178648.1 | - | 5 | 470 |