Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032664.3 | NP_116053.3 | 3 | 523 | ||
XM_011544676.2 | XP_011542978.1 | 4 | 995 | |||
XM_011544677.2 | XP_011542979.1 | 3 | 521 | |||
XM_011544678.2 | XP_011542980.1 | 4 | 528 | |||
XM_011544679.2 | XP_011542981.1 | 4 | 991 | |||
XM_011544680.2 | XP_011542982.1 | 4 | 989 | |||
XM_011544681.2 | XP_011542983.1 | 4 | 990 | |||
GenBank mRNA | AJ512465.1 | - | 3 | 511 | ||
AJ535838.1 | - | 4 | 561 | |||
AJ535839.1 | - | 3 | 501 | |||
AJ582015.1 | - | 3 | 523 | |||
AK098505.1 | - | 3 | 490 | |||
AK292993.1 | - | 3 | 521 | |||
AK297132.1 | - | 4 | 616 | |||
AK303231.1 | - | 3 | 456 | |||
BC004884.2 | - | 3 | 460 | |||
BC063462.1 | - | 3 | 563 | 5198060 |