Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033087.3 | NP_149078.1 | 2 | 880 | ||
NR_024532.1 | 3 | 1039 | ||||
GenBank mRNA | AB161356.1 | - | 2 | 837 | ||
AK027417.1 | - | 2 | 832 | |||
AK027814.1 | - | 1 | 1632 | |||
AK074704.1 | - | 2 | 832 | |||
AK074988.1 | - | 3 | 1017 | |||
AK075172.1 | - | 2 | 856 | |||
AK075189.1 | - | 1 | 100 | |||
AY358697.1 | - | 2 | 624 | |||
BC017876.1 | - | 2 | 830 | 4698763 |