Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033109.4 | NP_149100.2 | 28 | 2964 | ||
XM_005264629.2 | XP_005264686.1 | 28 | 2894 | |||
XM_017005172.1 | XP_016860661.1 | 27 | 2844 | |||
GenBank mRNA | BC053660.1 | - | 28 | 2879 | 6062060 | |
CR749867.1 | - | 9 | 1266 |