Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033261.2 | NP_150286.1 | 2 | 160 | ||
GenBank mRNA | AF271725.1 | - | 1 | 136 | ||
AK056950.1 | - | 2 | 160 | |||
BC017778.1 | - | 2 | 159 | 4291149 |